Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
1,3-Diethylurea
1,3-Diethylurea 97%
1,3-Diethylurea 97%
Bottle
APOSOR323264-25G
New Product
:
Apollo Scientific
1,3-Diethylurea 97%
APOSOR323264-25G
:
Apollo Scientific
New Product
:
623-76-7
1,3-Diethylurea 97%
Bottle
:
623-76-7
26,10 €
1 * 25 g
-
+
:
N,N'-Diethylurea
:
623-76-7
:
C₅H₁₂N₂O
:
MFCD00009028
:
116.16 g/mol
:
109…111 °C
:
Ambient
Pack type:
Bottle