Order Entry
Export
Orders LinkContactUsLinkComponent
Murexide indicator

Murexide indicator

Bottle
Catalog # APOSOR2362-100G
Irritant
New Product
Supplier:   Apollo Scientific
Murexide indicator
Catalog # APOSOR2362-100G
Supplier:  Apollo Scientific
New Product
CAS Number:  3051-09-0
Murexide indicator

Bottle

CAS Number:  3051-09-0
Price
34,80 €
Log in for availability
1 * 100 g
Product Details & Documents

Usage: Organic building block/reagent.

Synonyms:  Ammonium purpurate, Murexide triturate, Ammonium 5-(2,4,6-trioxoperhydropyrimidin-5-ylideneamino)barbiturate, Purpuric acid ammonium salt
  • CAS Number:3051-09-0
  • Formula:C₈H₅N₅O₆·H₃N
  • MDL Number:MFCD00012777
  • Molecular Weight:284,18572 g/mol
  • EINECS:221-266-6
Specifications

Specifications

  • Pack type:
    Bottle

Specifications