Order Entry
Export
Orders LinkContactUsLinkComponent
3'-Acetylacetophenone 98%

3'-Acetylacetophenone 98%

Bottle
Catalog # APOSOR59987-1G
New Product
Supplier:   Apollo Scientific
3'-Acetylacetophenone 98%
Catalog # APOSOR59987-1G
Supplier:  Apollo Scientific
New Product
CAS Number:  6781-42-6
3'-Acetylacetophenone 98%

Bottle

CAS Number:  6781-42-6
Price
21,80 €
Log in for availability
1 * 1 g
Product Details & Documents

Usage: Organic building block/reagent.

Synonyms:  Benzene-1,3-bis(acetyl)
  • CAS Number:6781-42-6
  • Formula:C₁₀H₁₀O₂
  • MDL Number:MFCD00008740
  • Molecular Weight:162,18519 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications