Order Entry
Export
Orders LinkContactUsLinkComponent
2-Mercaptopyrimidine

2-Mercaptopyrimidine

Bottle
Catalog # APOSOR29835-500G
Irritant
New Product
Supplier:   Apollo Scientific
2-Mercaptopyrimidine
Catalog # APOSOR29835-500G
Supplier:  Apollo Scientific
New Product
CAS Number:  1450-85-7
2-Mercaptopyrimidine

Bottle

CAS Number:  1450-85-7
Price
372,00 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  2-Thiopyrimidine
  • CAS Number:1450-85-7
  • Formula:C₄H₄N₂S
  • MDL Number:MFCD00006073
  • Molecular Weight:112.16 g/mol
  • Melting Pt:230 °C (dec.)
  • Storage Temperature:Ambient
  • EINECS:215-917-3
Specifications

Specifications

  • Pack type:
    Bottle

Specifications