4,4'-Dihydroxyazobenzene
Bottle
Catalog # APOSOR305565-250MG
New Product
Supplier: Apollo Scientific
4,4'-Dihydroxyazobenzene
Catalog # APOSOR305565-250MG
Supplier: Apollo Scientific
New Product
CAS Number: 2050-16-0
Jump to:
Product Details & Documents
Synonyms: 4,4-Dihydroxyazobenzene, 4,4'-Azodiphenol
- CAS Number:2050-16-0
- Formula:C₁₂H₁₀N₂O₂
- MDL Number:MFCD00045778
- Molecular Weight:214.22 g/mol
- Melting Pt:215 °C
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle



