4-Hydroxycinnamic acid 98%
Bottle
Catalog # APOSOR21362-500G
New Product
Supplier: Apollo Scientific
4-Hydroxycinnamic acid 98%
Catalog # APOSOR21362-500G
Supplier: Apollo Scientific
New Product
CAS Number: 7400-08-0
Jump to:
Product Details & Documents
Synonyms: p-Coumaric acid, p-Cumaric acid, 3-(4-Hydroxyphenyl)acrylic acid, p-Hydroxy-Cinnamic Acid
- CAS Number:7400-08-0
- Formula:C₉H₈O₃
- MDL Number:MFCD00004399
- Molecular Weight:164.16 g/mol
- Boiling Pt:346 °C
- Melting Pt:212…214 °C
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle



