1,9-Diaminononane
Bottle
Catalog # APOSOR10105-250MG
New Product
Supplier: Apollo Scientific
1,9-Diaminononane
Catalog # APOSOR10105-250MG
Supplier: Apollo Scientific
New Product
CAS Number: 646-24-2
Jump to:
Product Details & Documents
Synonyms: 1,9-Nonanediamine, Nonamethylendiamine
- CAS Number:646-24-2
- Formula:C₉H₂₂N₂
- MDL Number:MFCD00008251
- Molecular Weight:158.29 g/mol
- Boiling Pt:258…259 °C
- Melting Pt:35…40 °C
- Flash Pt:>110 °C
- Storage Temperature:Ambient
- EINECS:211-470-3
- UN:3259
- ADR:8,III
Specifications
Specifications
- Pack type:Bottle



