Order Entry
Export
Orders LinkContactUsLinkComponent
1,3-Diethylurea 97%

1,3-Diethylurea 97%

Bottle
Catalog # APOSOR323264-2.5KG
New Product
Supplier:   Apollo Scientific
1,3-Diethylurea 97%
Catalog # APOSOR323264-2.5KG
Supplier:  Apollo Scientific
New Product
CAS Number:  623-76-7
1,3-Diethylurea 97%

Bottle

CAS Number:  623-76-7
Price
1 720,00 €
Log in for availability
1 * 2,5 kg
Product Details & Documents
Synonyms:  N,N'-Diethylurea
  • CAS Number:623-76-7
  • Formula:C₅H₁₂N₂O
  • MDL Number:MFCD00009028
  • Molecular Weight:116.16 g/mol
  • Melting Pt:109…111 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications