Order Entry
Export
Orders LinkContactUsLinkComponent
 

Mannose triflate

Bottle
Catalog # APOSPC9900-25G
Irritant
New Product
Supplier:   Apollo Scientific
Mannose triflate
Catalog # APOSPC9900-25G
Supplier:  Apollo Scientific
New Product
CAS Number:  92051-23-5
Mannose triflate

Bottle

CAS Number:  92051-23-5
Price
2 720,00 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  (2S,3S,4S,5R,6R)-6-(Acetoxymethyl)-3-(((trifluoromethyl)sulphonyl)oxy)tetrahydro-2H-pyran-2,4,5-triyl triacetate, 1,3,4,6-Tetra-O-acetyl-2-O-(trifluoromethanesulphonyl)-β-D-mannopyranose, TATM
  • CAS Number:92051-23-5
  • Formula:C₁₅H₁₉F₃O₁₂S
  • MDL Number:MFCD00012353
  • Molecular Weight:480.36 g/mol
  • Melting Pt:118…122 °C
  • Storage Temperature:Freezer
Specifications

Specifications

  • Pack type:
    Bottle

Specifications