1,6-Hexanediol 97%
Bottle
Catalog # APOSOR322268-500G
New Product
Supplier: Apollo Scientific
1,6-Hexanediol 97%
Catalog # APOSOR322268-500G
Supplier: Apollo Scientific
New Product
CAS Number: 629-11-8
Jump to:
Product Details & Documents
Synonyms: Hexamethylene glycol, 1,6-Dihydroxy hexane
- CAS Number:629-11-8
- Formula:C₆H₁₄O₂
- MDL Number:MFCD00002985
- Molecular Weight:118.18 g/mol
- Boiling Pt:248…250 °C
- Melting Pt:40…43 °C
- Density:1.12 g/cm³ (25 °C)
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle



