D-Xylose 98%
Bottle
Catalog # APOSBIK9014-500G
New Product
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Usage: Biochemical/ life science applications.
- CAS Number:58-86-6
- Formula:C₅H₁₀O₅
- MDL Number:MFCD00151475
- Molecular Weight:150,1299 g/mol
Specifications
Specifications
- Pack type:Bottle



