Order Entry
Export
Orders LinkContactUsLinkComponent
3,5-Dinitrobenzonitrile 98+%

3,5-Dinitrobenzonitrile 98+%

Bottle
Catalog # APOSOR21915-1G
Irritant
New Product
Supplier:   Apollo Scientific
3,5-Dinitrobenzonitrile 98+%
Catalog # APOSOR21915-1G
Supplier:  Apollo Scientific
New Product
CAS Number:  4110-35-4
3,5-Dinitrobenzonitrile 98+%

Bottle

CAS Number:  4110-35-4
Price
21,80 €
Log in for availability
1 * 1 g
Product Details & Documents
  • CAS Number:4110-35-4
  • Formula:C₇H₃N₃O₄
  • MDL Number:MFCD00007228
  • Molecular Weight:193.12 g/mol
  • Melting Pt:126…130 °C
  • Storage Temperature:Ambient
  • EINECS:223-889-9
  • UN:3439
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Bottle

Specifications