ADA ≥98%
Bottle
Catalog # APOSBIA0302-2.5KG
New Product
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Biological buffer - useful range 6.0 - 7.2
Synonyms: 2,2'-[(2-Amino-2-oxoethyl)imino]diacetic acid, N-(2-Acetamido)iminodiacetic acid, 2,2'-((2-Amino-2-oxoethyl)azanediyl)diacetic acid
- CAS Number:26239-55-4
- Formula:H₂NC(O)CH₂N(CH₂COOH)₂
- MDL Number:MFCD00008031
- Molecular Weight:190.16 g/mol
- Melting Pt:219 °C
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle



