Order Entry
Export
Orders LinkContactUsLinkComponent
 

2,2-Diphenylethylamine 96%

Bottle
Catalog # APOSOR12742-500G
New Product
Supplier:   Apollo Scientific
2,2-Diphenylethylamine 96%
Catalog # APOSOR12742-500G
Supplier:  Apollo Scientific
New Product
CAS Number:  3963-62-0
2,2-Diphenylethylamine 96%

Bottle

CAS Number:  3963-62-0
Price
1 660,00 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  1-Amino-2,2-diphenylethane, 2,2-Diphenylethanamine
  • CAS Number:3963-62-0
  • Formula:C₁₄H₁₅N
  • MDL Number:MFCD00008143
  • Molecular Weight:197.28 g/mol
  • Melting Pt:48…49 °C
  • Flash Pt:113 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications