0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
Chromone
BP0000703969
Chromone
Bottle
APOSOR22414-500G
New Product
:
Apollo Scientific
Chromone
APOSOR22414-500G
:
Apollo Scientific
New Product
:
Chromone
Bottle
:
1 560,00 €
1 * 500 g
-
+
Pack type:
Bottle