Order Entry
Export
Orders LinkContactUsLinkComponent
 

2,4-Dimethoxybenzoic acid 98%

Bottle
Catalog # APOSOR345154-2.5KG
New Product
Supplier:   Apollo Scientific
2,4-Dimethoxybenzoic acid 98%
Catalog # APOSOR345154-2.5KG
Supplier:  Apollo Scientific
New Product
CAS Number:  91-52-1
2,4-Dimethoxybenzoic acid 98%

Bottle

CAS Number:  91-52-1
Price
1 760,00 €
Log in for availability
1 * 2,5 kg
Product Details & Documents

Usage: Organic building block/ reagent.

  • CAS Number:91-52-1
  • Formula:C₉H₁₀O₄
  • MDL Number:MFCD00002434
  • Molecular Weight:182,1733 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications