Order Entry
Export
Orders LinkContactUsLinkComponent
1,1'-Dibenzoylferrocene

1,1'-Dibenzoylferrocene

Bottle
Catalog # APOSOR315065-500G
New Product
Supplier:   Apollo Scientific
1,1'-Dibenzoylferrocene
Catalog # APOSOR315065-500G
Supplier:  Apollo Scientific
New Product
CAS Number:  12180-80-2
1,1'-Dibenzoylferrocene

Bottle

CAS Number:  12180-80-2
Price
1 490,00 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  Di(benzoylcyclopentadienyl) Iron, Bis(benzoylcyclopentadienyl) Iron
  • CAS Number:12180-80-2
  • Formula:C₂₄H₁₀FeO₂
  • MDL Number:MFCD00045455
  • Molecular Weight:386.18 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications