Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
trans-1,2-Cyclohexanedicarboxylic acid
trans-1,2-Cyclohexanedicarboxylic acid
trans-1,2-Cyclohexanedicarboxylic acid
Bottle
APOSOR350372-5G
New Product
:
Apollo Scientific
trans-1,2-Cyclohexanedicarboxylic acid
APOSOR350372-5G
:
Apollo Scientific
New Product
:
2305-32-0
trans-1,2-Cyclohexanedicarboxylic acid
Bottle
:
2305-32-0
21,80 €
1 * 5 g
-
+
:
trans-Cyclohexane-1,2-dicarboxylic acid
:
2305-32-0
:
C₈H₁₂O₄
:
MFCD00001464
:
172.18 g/mol
:
Ambient
Pack type:
Bottle