Order Entry
Export
Orders LinkContactUsLinkComponent
trans-1,2-Cyclohexanedicarboxylic acid

trans-1,2-Cyclohexanedicarboxylic acid

Bottle
Catalog # APOSOR350372-2.5KG
Irritant
New Product
Supplier:   Apollo Scientific
trans-1,2-Cyclohexanedicarboxylic acid
Catalog # APOSOR350372-2.5KG
Supplier:  Apollo Scientific
New Product
CAS Number:  2305-32-0
trans-1,2-Cyclohexanedicarboxylic acid

Bottle

CAS Number:  2305-32-0
Price
730,00 €
Log in for availability
1 * 2,5 kg
Product Details & Documents
Synonyms:  trans-Cyclohexane-1,2-dicarboxylic acid
  • CAS Number:2305-32-0
  • Formula:C₈H₁₂O₄
  • MDL Number:MFCD00001464
  • Molecular Weight:172.18 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications