0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
3,9-Dibromoperylene
BP0000737243
3,9-Dibromoperylene
3,9-Dibromoperylene
Bottle
APOSOR1027492-25G
New Product
:
Apollo Scientific
3,9-Dibromoperylene
APOSOR1027492-25G
:
Apollo Scientific
New Product
:
Expand image
3,9-Dibromoperylene
Bottle
Add to cart
:
1 820,00 €
Unit of Measure
1 * 25 g
-
+
Add to cart
Pack type:
Bottle