Cyclohexanone 2,4-Dinitrophenylhydrazone 99%
Bottle
Catalog # APOSOR1027545-25G
New Product
Supplier: Apollo Scientific
Cyclohexanone 2,4-Dinitrophenylhydrazone 99%
Catalog # APOSOR1027545-25G
Supplier: Apollo Scientific
New Product
CAS Number: 1589-62-4
Jump to:
Product Details & Documents
Usage: Organic building block/reagent
- CAS Number:1589-62-4
- Formula:C₁₂H₁₄N₄O₄
- Molecular Weight:278,26 g/mol
Specifications
Specifications
- Pack type:Bottle



