Order Entry
Export
Orders LinkContactUsLinkComponent
Cyclohexanone 2,4-Dinitrophenylhydrazone 99%

Cyclohexanone 2,4-Dinitrophenylhydrazone 99%

Bottle
Catalog # APOSOR1027545-1G
New Product
Supplier:   Apollo Scientific
Cyclohexanone 2,4-Dinitrophenylhydrazone 99%
Catalog # APOSOR1027545-1G
Supplier:  Apollo Scientific
New Product
CAS Number:  1589-62-4
Cyclohexanone 2,4-Dinitrophenylhydrazone 99%

Bottle

CAS Number:  1589-62-4
Price
138,00 €
Log in for availability
1 * 1 g
Product Details & Documents

Usage: Organic building block/reagent

  • CAS Number:1589-62-4
  • Formula:C₁₂H₁₄N₄O₄
  • Molecular Weight:278,26 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications