0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
9(10)-Dehydronandrolone
BP0000710371
9(10)-Dehydronandrolone
Bottle
APOSOR1027203-25G
New Product
:
Apollo Scientific
9(10)-Dehydronandrolone
APOSOR1027203-25G
:
Apollo Scientific
New Product
:
9(10)-Dehydronandrolone
Bottle
:
795,00 €
1 * 25 g
-
+
Pack type:
Bottle