Order Entry
Export
Orders LinkContactUsLinkComponent
9(10)-Dehydronandrolone 98%

9(10)-Dehydronandrolone 98%

Bottle
Catalog # APOSOR1027203-1G
New Product
Supplier:   Apollo Scientific
9(10)-Dehydronandrolone 98%
Catalog # APOSOR1027203-1G
Supplier:  Apollo Scientific
New Product
CAS Number:  
9(10)-Dehydronandrolone 98%

Bottle

CAS Number:  
Price
125,00 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  17beta-Hydroxyestra-4,9-dien-3-one
  • Molecular Weight:272,39 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications