Order Entry
Export
Orders LinkContactUsLinkComponent
 

Decoquinate 98%

Bottle
  APOSOR1030278-100G
New Product
Decoquinate 98%
  APOSOR1030278-100G
New Product
 :  18507-89-6
Decoquinate 98%

Bottle

 :  18507-89-6
 
155,00 €
   
1 * 100 g
 

Usage: Organic building block/reagent

 :  Ethyl 6-Decyloxy-7-ethoxy-4-hydroxyquinoline-3-carboxylate, Ethyl 6-(decyloxy)-7-ethoxy-4-hydroxyquinoline-3-carboxylate, 6-Decyloxy-7-ethoxy-4-hydroxyquinoline-3-carboxylic Acid Ethyl Ester
  •  :18507-89-6
  •  :C₂₄H₃₅NO₅
  •  :417,54 g/mol
 

 

  • Pack type:
    Bottle