L-Thyroxine 97%
Bottle
Catalog # APOSOR1030066-25G
New Product
Supplier: Apollo Scientific
L-Thyroxine 97%
Catalog # APOSOR1030066-25G
Supplier: Apollo Scientific
New Product
CAS Number: 51-48-9
Jump to:
Product Details & Documents
Usage: Organic building block/ reagent.
- Store at 2 to 8 °C
Synonyms: (S)-2-Amino-3-(4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl)propanoic acid
- CAS Number:51-48-9
- Formula:C₁₅H₁₁I₄NO₄
- MDL Number:MFCD00002595
- Molecular Weight:776,87 g/mol
- Storage Temperature:Refrigerator
- EINECS:200-101-1
Specifications
Specifications
- Pack type:Bottle



