Order Entry
Export
Orders LinkContactUsLinkComponent
L-Thyroxine 97%

L-Thyroxine 97%

Bottle
Catalog # APOSOR1030066-5G
Health
New Product
Supplier:   Apollo Scientific
L-Thyroxine 97%
Catalog # APOSOR1030066-5G
Supplier:  Apollo Scientific
New Product
CAS Number:  51-48-9
L-Thyroxine 97%

Bottle

CAS Number:  51-48-9
Price
49,20 €
Log in for availability
1 * 5 g
Product Details & Documents

Usage: Organic building block/ reagent.

  • Store at 2 to 8 °C
Synonyms:  (S)-2-Amino-3-(4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl)propanoic acid
  • CAS Number:51-48-9
  • Formula:C₁₅H₁₁I₄NO₄
  • MDL Number:MFCD00002595
  • Molecular Weight:776,87 g/mol
  • Storage Temperature:Refrigerator
  • EINECS:200-101-1
Specifications

Specifications

  • Pack type:
    Bottle

Specifications