Order Entry
Export
Orders LinkContactUsLinkComponent
 

1,2-Diphenylethane-1,2-Diol 98%

Bottle
Catalog # APOSOR1027818-25G
New Product
Supplier:   Apollo Scientific
1,2-Diphenylethane-1,2-Diol 98%
Catalog # APOSOR1027818-25G
Supplier:  Apollo Scientific
New Product
CAS Number:  492-70-6
1,2-Diphenylethane-1,2-Diol 98%

Bottle

CAS Number:  492-70-6
Price
1 890,00 €
Log in for availability
1 * 25 g
Product Details & Documents

Usage: Organic building block/reagent

  • CAS Number:492-70-6
  • Formula:C₁₄H₁₄O₂
  • Molecular Weight:214,26 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications