Order Entry
Export
Orders LinkContactUsLinkComponent
Solvent Blue 38

Solvent Blue 38

Bottle
Catalog # APOSIN12079-2.5KG
New Product
Supplier:   Apollo Scientific
Solvent Blue 38
Catalog # APOSIN12079-2.5KG
Supplier:  Apollo Scientific
New Product
CAS Number:  1328-51-4
Solvent Blue 38

Bottle

CAS Number:  1328-51-4
Price
1 380,00 €
Log in for availability
1 * 2,5 kg
Product Details & Documents

Usage: Inorganic salt or catalyst.

Synonyms:  Direct Blue 86, C.I. 74180, Luxol® Fast Blue MBSN
  • CAS Number:1328-51-4
  • Formula:C₃₂H₁₂CuN₈Na₂O₆S₂
  • Molecular Weight:778,14 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications