Order Entry
Export
Orders LinkContactUsLinkComponent
AC-55541 98%

AC-55541 98%

Bottle
Catalog # APOSOR1026543-250M
New Product
Supplier:   Apollo Scientific
AC-55541 98%
Catalog # APOSOR1026543-250M
Supplier:  Apollo Scientific
New Product
CAS Number:  916170-19-9
AC-55541 98%

Bottle

CAS Number:  916170-19-9
Price
400,00 €
Log in for availability
1 * 250 mg
Product Details & Documents

Usage: Organic building block/reagent

Synonyms:  (E)-N-(2-(2-(1-(3-Bromophenyl)ethylidene)hydrazineyl)-2-oxo-1-(4-oxo-3,4-dihydrophthalazin-1-yl)ethyl)benzamide
  • CAS Number:916170-19-9
  • Formula:C₂₅H₂₀BrN₅O₃
  • Molecular Weight:518,36 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications