Order Entry
Export
Orders LinkContactUsLinkComponent
Fast red ITR base 98%

Fast red ITR base 98%

Bottle
Catalog # APOSOR1029080-100G
New Product
Supplier:   Apollo Scientific
Fast red ITR base 98%
Catalog # APOSOR1029080-100G
Supplier:  Apollo Scientific
New Product
CAS Number:  97-35-8
Fast red ITR base 98%

Bottle

CAS Number:  97-35-8
Price
34,80 €
Log in for availability
1 * 100 g
Product Details & Documents

Usage: Organic building block/ reagent.

  • Non-hazardous
Synonyms:  Azoic Diazo Component 42, N1,N1-Diethyl-4-methoxymetanilamide, Fast Red ITR, 3-Amino-N,N-diethyl-4-methoxybenzenesulphonamide
  • CAS Number:97-35-8
  • Formula:C₁₁H₁₈N₂O₃S
  • Molecular Weight:258,34 g/mol
  • EINECS:202-575-5
Specifications

Specifications

  • Pack type:
    Bottle

Specifications