Giemsa stain biological stain
Bottle
Catalog # APOSOR1028634-2.5K
New Product
Supplier: Apollo Scientific
Giemsa stain biological stain
Catalog # APOSOR1028634-2.5K
Supplier: Apollo Scientific
New Product
CAS Number: 51811-82-6
Jump to:
Product Details & Documents
Usage: Organic building block/reagent
Synonyms: Azure mixture sicc. Giemsa stain, Azur-Eosin-Methylene blue acc. Giemsa, Giemsa's azur-eosin-methylene blue
- CAS Number:51811-82-6
- Formula:C₁₄Cl₁H₁₄N₃S₁
- Molecular Weight:291,8 g/mol
Specifications
Specifications
- Pack type:Bottle
- Cat. No.:APOSOR1028634-2.5K



