2,2-Dimethoxybiphenyl 97%
Bottle
Catalog # APOSOR1028428-500G
New Product
Supplier: Apollo Scientific
2,2-Dimethoxybiphenyl 97%
Catalog # APOSOR1028428-500G
Supplier: Apollo Scientific
New Product
CAS Number: 4877-93-4
Jump to:
Product Details & Documents
Usage: Organic building block/ reagent.
- CAS Number:4877-93-4
- Formula:C₁₄H₁₄O₂
- Molecular Weight:214,26 g/mol
Specifications
Specifications
- Pack type:Bottle



