Order Entry
Export
Orders LinkContactUsLinkComponent
(±)-10-Hydroxycamptothecin ≥98%

(±)-10-Hydroxycamptothecin ≥98%

Bottle
Catalog # APOSBUP11318-500MG
New Product
Supplier:   Apollo Scientific
(±)-10-Hydroxycamptothecin ≥98%
Catalog # APOSBUP11318-500MG
Supplier:  Apollo Scientific
New Product
CAS Number:  64439-81-2
(±)-10-Hydroxycamptothecin ≥98%

Bottle

CAS Number:  64439-81-2
Price
361,00 €
Log in for availability
1 * 500 mg
Product Details & Documents

Usage: Biochemical/life science applications.

Synonyms:  1-HCPT, 10-Hydroxycamptothecin, 4-Ethyl-4,9-dihydroxy-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione, NSC107124
  • CAS Number:64439-81-2
  • Formula:C₂₀H₁₆N₂O₅
  • Molecular Weight:364,35 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications