Order Entry
Export
Orders LinkContactUsLinkComponent
Bromochlorophenolblue 95% (dye content)

Bromochlorophenolblue 95% (dye content)

Bottle
Catalog # APOSOR1028099-5G
New Product
Supplier:   Apollo Scientific
Bromochlorophenolblue 95% (dye content)
Catalog # APOSOR1028099-5G
Supplier:  Apollo Scientific
New Product
CAS Number:  2553-71-1
Bromochlorophenolblue 95% (dye content)

Bottle

CAS Number:  2553-71-1
Price
246,00 €
Log in for availability
1 * 5 g
Product Details & Documents

Usage: Organic building block/reagent

Synonyms:  4,4'-(3H-2,1-Benzoxathiol-3-ylidene)bis[2-bromo-6-chlorophenol] S,S-dioxide
  • CAS Number:2553-71-1
  • Formula:C₁₉H₁₀Br₂Cl₂O₅S
  • Molecular Weight:581,06 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications