Order Entry
Export
Orders LinkContactUsLinkComponent
 

Antitubercular agent-30 ≥98%

Bottle
Catalog # APOSBUP10953-200MG
New Product
Supplier:   Apollo Scientific
Antitubercular agent-30 ≥98%
Catalog # APOSBUP10953-200MG
Supplier:  Apollo Scientific
New Product
CAS Number:  384857-54-9
Antitubercular agent-30 ≥98%

Bottle

CAS Number:  384857-54-9
Price
620,00 €
Log in for availability
1 * 200 mg
Product Details & Documents

Usage: Biochemical/ life science applications.

Synonyms:  Antitubercular agent-30
  • CAS Number:384857-54-9
  • Formula:C₁₂H₁₀N₂O₃S
  • Molecular Weight:262,28 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications