0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
10-Nonadecanone
Nonadecan-10-one 98%
Nonadecan-10-one 98%
Bottle
APOSOR95811-1G
New Product
:
Apollo Scientific
Nonadecan-10-one 98%
APOSOR95811-1G
:
Apollo Scientific
New Product
:
504-57-4
Nonadecan-10-one 98%
Bottle
:
504-57-4
21,80 €
1 * 1 g
-
+
Usage: Organic building block/reagent
:
Caprinone, Dinonyl ketone
:
504-57-4
:
C₁₉H₃₈O
:
MFCD00009584
:
282,5 g/mol
Pack type:
Bottle