Order Entry
Export
Orders LinkContactUsLinkComponent
Benzophenoneoxime 98%

Benzophenoneoxime 98%

Bottle
Catalog # APOSOR75811-500G
New Product
Supplier:   Apollo Scientific
Benzophenoneoxime 98%
Catalog # APOSOR75811-500G
Supplier:  Apollo Scientific
New Product
CAS Number:  574-66-3
Benzophenoneoxime 98%

Bottle

CAS Number:  574-66-3
Price
686,00 €
Log in for availability
1 * 500 g
Product Details & Documents

Usage: Organic building block/ reagent.

  • CAS Number:574-66-3
  • Formula:C₁₃H₁₁NO
  • MDL Number:MFCD00051461
  • Molecular Weight:197,23 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications