Order Entry
Export
Orders LinkContactUsLinkComponent
Human recombinant BD-2

Human recombinant BD-2

1 mg
Catalog # APOSBITP2072-1MG
New Product
Supplier:   Apollo Scientific
Human recombinant BD-2
Catalog # APOSBITP2072-1MG
Supplier:  Apollo Scientific
New Product
CAS Number:  
Human recombinant BD-2

1 mg

CAS Number:  
Price
3 500,00 €
Log in for availability
1 * 1 mg
Product Details & Documents
Specifications

Specifications

  • Pk:
    1 mg
  • Cat. No.:
    APOSBITP2072-1MG
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    BD-2
  • Protein synonyms:
    Beta Defensin -2
  • Species:
    Human

Specifications