1,9-Dichlorononane 98%
Bottle
Catalog # APOSOR80225-5G
New Product
Supplier: Apollo Scientific
1,9-Dichlorononane 98%
Catalog # APOSOR80225-5G
Supplier: Apollo Scientific
New Product
CAS Number: 821-99-8
Jump to:
Product Details & Documents
Usage: Organic building block/ reagent.
- CAS Number:821-99-8
- Formula:C₉H₁₈Cl₂
- MDL Number:MFCD00001024
- Molecular Weight:197,15 g/mol
Specifications
Specifications
- Pack type:Bottle



