0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
2,6-Diethylaniline
BP0000757150
2,6-Diethylaniline
2,6-Diethylaniline
Bottle
APOSOR93652-100G
New Product
:
Apollo Scientific
2,6-Diethylaniline
APOSOR93652-100G
:
Apollo Scientific
New Product
:
Expand image
2,6-Diethylaniline
Bottle
Add to cart
:
24,60 €
Unit of Measure
1 * 100 g
-
+
Add to cart
Pack type:
Bottle