4,7-Dibromoquinoline
Bottle
Catalog # APOSOR307502-100MG
New Product
Supplier: Apollo Scientific
4,7-Dibromoquinoline
Catalog # APOSOR307502-100MG
Supplier: Apollo Scientific
New Product
CAS Number: 700871-88-1
Jump to:
Product Details & Documents
Synonyms: 4,7-Dibromo-1-azanaphthalene
- CAS Number:700871-88-1
- Formula:C₉H₅Br₂N
- MDL Number:MFCD08063188
- Molecular Weight:286.951 g/mol
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle



