Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
1,4-Oxazepane
1,4-Oxazepane
1,4-Oxazepane
Bottle
APOSOR322438-5G
New Product
:
Apollo Scientific
1,4-Oxazepane
APOSOR322438-5G
:
Apollo Scientific
New Product
:
5638-60-8
1,4-Oxazepane
Bottle
:
5638-60-8
316,00 €
1 * 5 g
-
+
:
Homomorpholine
:
5638-60-8
:
C₅H₁₁NO
:
MFCD09971804
:
101.1 g/mol
:
Ambient
Pack type:
Bottle