Order Entry
Export
Orders LinkContactUsLinkComponent
1,4-Oxazepane

1,4-Oxazepane

Bottle
Catalog # APOSOR322438-1G
New Product
Supplier:   Apollo Scientific
1,4-Oxazepane
Catalog # APOSOR322438-1G
Supplier:  Apollo Scientific
New Product
CAS Number:  5638-60-8
1,4-Oxazepane

Bottle

CAS Number:  5638-60-8
Price
65,20 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  Homomorpholine
  • CAS Number:5638-60-8
  • Formula:C₅H₁₁NO
  • MDL Number:MFCD09971804
  • Molecular Weight:101.1 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications