Order Entry
Export
Orders LinkContactUsLinkComponent
Galactitol

Galactitol

Bottle
Catalog # APOSBIB6049-25G
New Product
Supplier:   Apollo Scientific
Galactitol
Catalog # APOSBIB6049-25G
Supplier:  Apollo Scientific
New Product
CAS Number:  608-66-2
Galactitol

Bottle

CAS Number:  608-66-2
Price
37,70 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  Dulcitol, rel-(2R,3S,4R,5S)-Hexane-1,2,3,4,5,6-hexaol
  • CAS Number:608-66-2
  • Formula:HOH₂C(CH(OH))₄CH₂OH
  • MDL Number:MFCD00064288
  • Molecular Weight:182.17 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications