Order Entry
Export
Orders LinkContactUsLinkComponent
 

PD 98059

Bottle
Catalog # APOSBIP0711-1G
Irritant
New Product
Supplier:   Apollo Scientific
PD 98059
Catalog # APOSBIP0711-1G
Supplier:  Apollo Scientific
New Product
CAS Number:  167869-21-8
PD 98059

Bottle

CAS Number:  167869-21-8
Price
2 060,00 €
Log in for availability
1 * 1 g
Product Details & Documents

Specific inhibitor of MAPK kinase

Synonyms:  PD98059, 2'-Amino-3'-methoxyflavone, 2-(2-Amino-3-methoxyphenyl)chromone, PD 98059, 2-(2-Amino-3-methoxyphenyl)-4H-1-benzopyran-4-one, 2-(2-Amino-3-methoxyphenyl)-4H-chromen-4-one
  • CAS Number:167869-21-8
  • Formula:C₁₆H₁₃NO₃
  • MDL Number:MFCD00671789
  • Molecular Weight:267.284 g/mol
  • Storage Temperature:Freezer
Specifications

Specifications

  • Pack type:
    Bottle

Specifications