Order Entry
Export
Orders LinkContactUsLinkComponent
 

Fluorescein indicator

Bottle
Catalog # APOSOR1013318-500G
Irritant
New Product
Supplier:   Apollo Scientific
Fluorescein indicator
Catalog # APOSOR1013318-500G
Supplier:  Apollo Scientific
New Product
CAS Number:  2321-07-5
Fluorescein indicator

Bottle

CAS Number:  2321-07-5
Price
65,20 €
Log in for availability
1 * 500 g
Product Details & Documents

Usage: Organic building block/ reagent.

Synonyms:  3',6'-Dihydroxy-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one, 2-(6-Hydroxy-3-oxo-(3H)-xanthen-9-yl)benzoic acid
  • CAS Number:2321-07-5
  • Formula:C₂₀H₁₂O₅
  • MDL Number:MFCD00005050
  • Molecular Weight:332,31 g/mol
  • EINECS:219-031-8
Specifications

Specifications

  • Pack type:
    Bottle

Specifications