Order Entry
Export
Orders LinkContactUsLinkComponent
 

Perylene 98%

Bottle
Catalog # APOSOR1014604-500G
Irritant
New Product
Supplier:   Apollo Scientific
Perylene 98%
Catalog # APOSOR1014604-500G
Supplier:  Apollo Scientific
New Product
CAS Number:  198-55-0
Perylene 98%

Bottle

CAS Number:  198-55-0
Price
974,00 €
Log in for availability
1 * 500 g
Product Details & Documents

Usage: Organic building block/ reagent.

  • CAS Number:198-55-0
  • Formula:C₂₀H₁₂
  • MDL Number:MFCD00004142
  • Molecular Weight:252,31 g/mol
  • EINECS:205-900-9
Specifications

Specifications

  • Pack type:
    Bottle

Specifications