Perylene 98%
Bottle
APOSOR1014604-5G
New Product
Jump to:
Usage: Organic building block/ reagent.
- CAS Number:198-55-0
- Formula:C₂₀H₁₂
- MDL Number:MFCD00004142
- Molecular Weight:252,31 g/mol
- EINECS:205-900-9
Specifications
Specifications
- Pack type:Bottle



