Order Entry
Export
Orders LinkContactUsLinkComponent
 

Mouse Recombinant IL-19

1 mg
Catalog # APOSBITP1182-1MG
New Product
Supplier:   Apollo Scientific
Mouse Recombinant IL-19
Catalog # APOSBITP1182-1MG
Supplier:  Apollo Scientific
New Product
CAS Number:  
Mouse Recombinant IL-19

1 mg

CAS Number:  
Price
6 080,00 €
Log in for availability
1 * 1 mg
Product Details & Documents
Specifications

Specifications

  • Pk:
    1 mg
  • Cat. No.:
    APOSBITP1182-1MG
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    IL-19
  • Protein synonyms:
    Interleukin-19
  • Species:
    Mouse

Specifications